Key Documents
CLS5840250
Pyrex® volumetric flask, phosphoric acid
capacity 250 mL
Synonyme(s) :
flasks glass, volumetric flasks, volumetric glass, Corning® 5840
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
wide-neck
Stérilité
non-sterile
Caractéristiques
closure type ST stopper
Conditionnement
case of 1
Fabricant/nom de marque
Corning 5840-250
tol. measuring range
0.18 mL
Capacité
250 mL
Description générale
For use in determining phosphoric acid in mixed fertilizer. Of heavy construction throughout and with a wide neck.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.