Principaux documents
CLS5650100
Pyrex® volumetric flask, with screw cap
capacity 100 mL
Synonyme(s) :
flasks glass, volumetric flasks, volumetric glass, Corning® 5650
12 EA
784,00 $
784,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
12 EA
784,00 $
About This Item
Code UNSPSC :
41121804
784,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
borosilicate glass
long
Stérilité
non-sterile
Caractéristiques
closure type phenolic-style
Conditionnement
case of 12
pack of 6
Fabricant/nom de marque
Corning 5650-100
tol. measuring range
0.08 mL
Capacité
100 mL
Description générale
These standard design flasks conform to Class A capacity tolerances prescribed by ASTM. However, to facilitate transfer of crystals, salts and powders, the open end has been expanded on the 25 mL and 50 mL sizes. The standard 18-410 G.P.I screw caps have a durable cone-shaped polyethylene liner.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.