Principaux documents
CLS53411L
Pyrex® heavy-wall filtering flask with replaceable PP tubulations
capacity 1,000 mL
Synonyme(s) :
conical flasks, filtering flask, flasks erlenmeyer
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
narrow-mouth
Taille de l'obturateur
8
Stérilité
non-sterile
Caractéristiques
closure type Rubber stopper
Conditionnement
case of 12
pack of 6
Fabricant/nom de marque
Corning 5341-1L
Capacité
1,000 mL
Description générale
Permanent, white enamel graduations and marking spots and replaceable PP tubulation with neoprene grommet. Tubulation is 12.5 mm o.d.
Informations légales
PYREX is a registered trademark of Corning, Inc.