Principaux documents
CLS53404L
Pyrex® heavy wall graduated filtering flask, with tubulation
12.5 mm O.D. hose-connection, capacity 4,000 mL
Synonyme(s) :
conical flasks, filtering flasks, flasks erlenmeyer, Corning® 5340
1 EA
354,00 $
6 EA
1 630,00 $
354,00 $
Disponible pour expédition le16 avril 2025Détails
Changer de vue
1 EA
354,00 $
6 EA
1 630,00 $
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
354,00 $
Disponible pour expédition le16 avril 2025Détails
Matériaux
borosilicate glass
flat bottom flask
narrow-mouth
Joint
joint stopper12
Stérilité
non-sterile
Caractéristiques
closure type Rubber stopper
Conditionnement
pack of 1
case of 6
Fabricant/nom de marque
Corning 5340-4L
Plage de mesure du volume
1,500-4,000 mL
Capacité
4,000 mL
Description générale
These flasks have sidearm tabulations and are blown in special molds designed to give maximum mechanical strength. For convenience, the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.