Principaux documents
CLS44252XL
Pyrex® Fernbach baffled culture flask
with GL45 screw cap
Synonyme(s) :
Corning container
3 EA
900,00 $
900,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
3 EA
900,00 $
About This Item
Code UNSPSC :
41122104
Nomenclature NACRES :
NB.43
900,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
borosilicate glass
flat bottom flask
polypropylene cap
wide-mouth
wide-mouth neck
Stérilité
non-sterile
Caractéristiques
cap
closure type Screw cap
Conditionnement
case of 3
Fabricant/nom de marque
Corning 4425-2XL
Capacité
2,800 mL
Raccords
GL 45 neck
Description générale
These 2800mL PYREX® Fernbach-style culture flasks have triple baffles located at the center of the flask bottom to achieve maximal oxygen transfer to culture medium. These flasks have a large marking spot.
Note: Do not use this flask on a heat source smaller than the base of the flask. Uneven heating of the flask could cause breakage. They also have a single size linerless, one-piece autoclavable orange polypropylene plug seal caps with GL45 threads and drip-free pouring rings.
A wide range of optional caps are available for these bottles including:
  • Polypropylene colored caps for ease of sorting and identification
  • Vented membrane cap with 0.22μm PTFE hydrophobic membrane
  • PBT (polybutylene terephthalate) solid caps and ETFE pouring rings for dry heat sterilization (180°C).
  • PBT caps with silicone septa for syringe sampling or introduction of reagents.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.