Principaux documents
BR81146
BRAND® burette lateral stopcock
for burette capacity 25 mL, with spare key, glass key
Synonyme(s) :
Accessories, Burette stopcock, Burettes, Stopcocks
About This Item
Code UNSPSC :
41121808
Nomenclature NACRES :
NB.43
Matériaux
amber
glass key
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 81146
Description générale
BRAND burettes are always calibrated to deliver (TD, Ex).
Waiting times:
BLAUBRAND® - waiting time 30 sec. SILBERBRAND - no waiting time
A waiting time is usually not necessary if a titration is performed (see DIN EN ISO 385).
All BLAUBRAND® burettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
Informations légales
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG