BR4600221
BRAND® Dispensette® S Fixed-volume bottle-top dispenser
volume 2 mL, with recirculation valve
Synonyme(s) :
Fixed-Volume Dispenser
872,00 $
About This Item
872,00 $
Produits recommandés
Caractéristiques
DE-M marking
with recirculation valve
Fabricant/nom de marque
BRAND 4600221
Volume
2 mL
Raccords
(GL 28/S28)
GL 32
GL 38
S 40
Description générale
Caractéristiques et avantages
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
Contenu apparenté
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.