Principaux documents
BR144640
BRAND® BISTABIL® ground glass stopper, conical joint
joint: ST/NS 34/35
Synonyme(s) :
BRAND® BISTABIL® ground glass stopper conical joint stopper, Conical joint stoppers, Stoppers
1 EA
32,00 $
32,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
32,00 $
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.51
32,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Joint
joint: ST/NS34/35
Description
hexagonal grip
hollow
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 144640
Description générale
Conical ground joint stopper made from Duran®. Hollow with drip tip and hexagonal grip. Meets DIN 12 252.
DIN 12252. Hollow, with drip tip and hexagonal grip.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Bistabil is a registered trademark of Lenz GmbH
DURAN is a registered trademark of DWK Life Sciences