Principaux documents
BR144520
BRAND® conical joint stopper, borosilicate glass
joint: ST/NS 14/23, solid
Synonyme(s) :
Conical joint stoppers, Stoppers
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.51
Matériaux
glass
octagonal borosilicate glass (grip)
Joint
joint: ST/NS14/23
Description
octagonal grip
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 144520
Description générale
DIN 12252. Solid or semi-hollow, with octagonal grip.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG