Principaux documents
BR113135
BRAND® forcep with rectangular ends
PTFE, L 200 mm
Synonyme(s) :
Forceps, PTFE
1 EA
136,00 $
136,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
136,00 $
About This Item
Code UNSPSC :
41122405
Nomenclature NACRES :
NB.13
136,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
PTFE
Fabricant/nom de marque
BRAND 113135
Longueur
200 mm
Description générale
The BRAND® Forceps with Rectangular Ends are specialized tools designed for handling materials in challenging laboratory environments. Made from high-quality PTFE (polytetrafluoroethylene), these forceps offer exceptional durability and performance. The PTFE construction ensures excellent resistance to a wide range of chemicals, making these forceps suitable for use in corrosive environments. Capable of withstanding high temperatures, these forceps are ideal for applications that involve heat exposure.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG