Principaux documents
BMSV0020F1
Sigma-Aldrich
Aspire Laboratory Aspirator Accessories
Hydrophobic filter
1 EA
48,90 $
48,90 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
48,90 $
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.79
48,90 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pk of 1 ea (1 filter each included)
Fabricant/nom de marque
Benchmark Scientific V0020-F1
Description générale
Accessories designed for use with Aspire laboratory aspirator.
Aspire Laboratory Aspirator Accessories, Hydrophobic filter blocks the entering of fluids into the pump unit. It also protects the pump unit from bacterial contamination.
Caractéristiques et avantages
  • Provides a barrier to protect against bacterial contamination of pump unit
Informations légales
6538 is a trademark of American Type Culture Collection
Aspire is a trademark of Benchmark Scientific, Inc.