Documents
BMSV10028
Sigma-Aldrich
Aspire Laboratory Aspirator Accessories
Optional eight channel adapter, for use with 200 uL pipette tips
Synonyme(s) :
multichannel aspiration adapter, vacuum aspirator multichannel adapter
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.79
Fabricant/nom de marque
Benchmark Scientific V1002-8
Informations légales
6538 is a trademark of American Type Culture Collection