Principaux documents
BMSBR5000
Sigma-Aldrich
Everlast rocker with flat mat
AC/DC input 115 V AC, US 3-pin plug
1 EA
1 790,00 $
1 790,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
1 790,00 $
About This Item
Code UNSPSC :
41103811
Nomenclature NACRES :
NB.73
1 790,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Fabricant/nom de marque
Benchmark Scientific BR5000
Larg. × P
16 in. × 12 in.
Description générale
  • Stainless-steel platform
  • Dual platform accessory doubles work surface
  • Cold room and incubator safe
The rocking speed and tilt angle can easily be adjusted
to vary between gentle and aggressive agitation. Both
the standard platform and the (optional) stacking
platform are constructed of stainless steel, preventing
against any rusting or corrosion. The Everlast 247 is
designed to provide years of uninterrupted service.
Informations légales
Everlast is a trademark of Benchmark Scientific, Inc.