MilliporeSigma
Toutes les photos(5)
Principaux documents
BAF378741001
SP Bel-Art® Sterileware Powder Thief
capacity × L 1 mL (0.03 oz) × 39.375 in. (100 cm), pack of 20 ea (individually wrapped)
Synonyme(s) :
sampler
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
Matériaux
high-density polyethylene
Conditionnement
pack of 20 ea (individually wrapped)
Fabricant/nom de marque
Bel-Art F37874-1001
capacity × L
1 mL (0.03 oz) × 39.375 in. (100 cm)
Description générale
Available in two lengths each with four tip volumes, these sterile powder thieves easily sample from targeted locations within bulk powders. Used by pharmaceutical, food, cosmetic and chemical companies.
Caractéristiques et avantages
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) thief heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10^-6)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing
Note: Sterilized products are not returnable.
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) thief heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10^-6)Ready for one time use and disposal
  • Lot stamped for accurate tracing
Note: Sterilized products are not returnable.
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP