Principaux documents
BAF183171000
SP Bel-Art® Electrode Holder clip
for holding an electrode, a reference electrode and an ATC probe
Synonyme(s) :
electrode holder
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F18317-1000
Description générale
Flexible arm with molded acetal copolymer links has ball-and-socket joints. One end is designed to accept the F18317-1000 Electrode Holder (sold separately) while the other has a 1.3cm (½") diameter stainless steel pin for attachment to the weighted base or a suitable clamp.
Caractéristiques et avantages
  • Flexible arm with molded acetal copolymer links has ball-and-socket joints. One end is designed to accept the F18317-1000 Electrode Holder (sold separately) while the other has a 1.3cm (½") diameter stainless steel pin for attachment to the weighted base or a suitable clamp.
  • Octagonal-shaped wells (4 total) are centered along each edge of the base with a maximum diameter of 1.5cm (?") for affixing flexible arms, additional support rods, or holders (two thumb screws are provided)
  • Electrode holder (sold separately) snaps onto the end of the Flexible Gooseneck Arm affording 3 stepped wells for holding an electrode, a reference electrode and an ATC probe
  • Flexible arm measures 46cm (18") long x 2.5cm (1") in diameter
  • Sturdy base measures 23 x 16cm (9 x 6") and weighs 1.5kg (3.3lb)
  • Center well of the holder has an 1.8cm (¾") I.D., side wells each have a 1.7cm (?") I.D.; through holes are all 1.3cm (½") I.D.
  • Includes three reducing bushings for smaller diameters and five nylon cable ties for attaching wires to the arm
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP