Skip to Content
MilliporeSigma
All Photos(1)

Documents

V800224

Sigma-Aldrich

2-Methyl-1-propanol

LR, ≥99%

Synonym(s):

Isobutanol, Isobutyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2OH
CAS Number:
Molecular Weight:
74.12
Beilstein:
1730878
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:

grade

LR

vapor density

2.55 (vs air)

vapor pressure

8 mmHg ( 20 °C)
8.8 mmHg ( 0 °C)

product line

Vetec

Assay

≥99%

form

liquid

autoignition temp.

801 °F

expl. lim.

10.6 %

refractive index

n20/D 1.396 (lit.)

bp

108 °C (lit.)

mp

−108 °C (lit.)

density

0.803 g/mL at 25 °C (lit.)

SMILES string

CC(C)CO

InChI

1S/C4H10O/c1-4(2)3-5/h4-5H,3H2,1-2H3

InChI key

ZXEKIIBDNHEJCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arul M Varman et al.
Applied and environmental microbiology, 79(3), 908-914 (2012-11-28)
Global warming and decreasing fossil fuel reserves have prompted great interest in the synthesis of advanced biofuels from renewable resources. In an effort to address these concerns, we performed metabolic engineering of the cyanobacterium Synechocystis sp. strain PCC 6803 to
Dawid Brat et al.
FEMS yeast research, 13(2), 241-244 (2013-01-03)
Simultaneous overexpression of an optimized, cytosolically localized valine biosynthesis pathway together with overexpression of xylose isomerase XylA from Clostridium phytofermentans, transaldolase Tal1 and xylulokinase Xks1 enabled recombinant Saccharomyces cerevisiae cells to complement the valine auxotrophy of ilv2,3,5 triple deletion mutants
Jan-Karl Guterl et al.
ChemSusChem, 5(11), 2165-2172 (2012-10-23)
The limited supply of fossil resources demands the development of renewable alternatives to petroleum-based products. Here, biobased higher alcohols such as isobutanol are versatile platform molecules for the synthesis of chemical commodities and fuels. Currently, their fermentation-based production is limited
Genny A Pang et al.
The journal of physical chemistry. A, 116(19), 4720-4725 (2012-04-21)
This work presents the first direct experimental study of the rate constant for the reaction of OH with iso-butanol (2-methyl-1-propanol) at temperatures from 907 to 1147 K at near-atmospheric pressures. OH time-histories were measured behind reflected shock waves using a
Han Li et al.
Science (New York, N.Y.), 335(6076), 1596-1596 (2012-03-31)
One of the major challenges in using electrical energy is the efficiency in its storage. Current methods, such as chemical batteries, hydraulic pumping, and water splitting, suffer from low energy density or incompatibility with current transportation infrastructure. Here, we report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service