2-Methyl-1-propanol
Synonym(s):
Isobutanol, Isobutyl alcohol, 2-Methyl-1-propanol
Linear Formula:
(CH3)2CHCH2OH
CAS Number:
Molecular Weight:
74.12
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-9 of 9
Compare
Product Number
Description
Pricing