Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAAR5822700

Sodium diphenylamine-4-sulfonate

uniLAB

Synonym(s):

4-(Phenylamino)benzenesulfonic acid sodium salt, Diphenylamine-4-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10NNaO3S
CAS Number:
Molecular Weight:
271.27
MDL number:
UNSPSC Code:
12352116

product line

uniLAB

packaging

pkg of 10 g (SAAR5822700BX)

storage temp.

2-30°C

SMILES string

[Na+].[O-]S(=O)(=O)c1ccc(Nc2ccccc2)cc1

InChI

1S/C12H11NO3S.Na/c14-17(15,16)12-8-6-11(7-9-12)13-10-4-2-1-3-5-10;/h1-9,13H,(H,14,15,16);/q;+1/p-1

InChI key

DGXTZMPQSMIFEC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Chemically pure
For laboratory use only

Legal Information

uniLAB is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service