Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAAR5046200

Potassium oxalate monohydrate

univAR

Synonym(s):

Ethanedioic acid, Oxalic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(COOK)2 · H2O
CAS Number:
Molecular Weight:
184.23
Beilstein:
3752576
MDL number:

product line

univAR

packaging

poly bottle of 12 × 500 g (SAAR5046200EM)

storage temp.

2-30°C

SMILES string

O.[K+].[K+].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.2K.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);;;1H2/q;2*+1;/p-2

InChI key

QCPTVXCMROGZOL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service