Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAAR3223400

Iodine Indicator

Vitex

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116100

packaging

pkg of 100 g (SAAR3223400DC)

storage temp.

2-30°C

Application

Chemically pure
For laboratory use only

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service