Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09175

Supelco

pH-indicator solution pH 4.0 - 10.0

Universal Indicator with colour card pH 4.0 - 4.5 - 5.0 - 5.5 - 6.0 - 6.5 - 7.0 - 7.5 - 8.0 - 8.5 - 9.0 - 9.5 - 10.0

Synonym(s):

Indicator solutions universal, Universal indicator solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113034
NACRES:
NB.21

product name

pH-indicator solution pH 4.0 - 10.0, Universal Indicator with colour card pH 4.0 - 4.5 - 5.0 - 5.5 - 6.0 - 6.5 - 7.0 - 7.5 - 8.0 - 8.5 - 9.0 - 9.5 - 10.0

Quality Level

form

liquid

pH

5.5-6.0 (20 °C in H2O)

transition temp

flash point 23 °C

density

0.89 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

Application test: conforms

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service