Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAAR2236050

EDTA disodium salt solution

0.1 M (0.2 N)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107
Pricing and availability is not currently available.

packaging

glass bottle of 1 × 2.5 L (SAAR2236050LC)

concentration

0.1 M (0.2 N)

Application

For laboratory use only.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service