Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAAR191122

Potassium iodate-iodide N/64 solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302

packaging

glass bottle of 4 × 2.5 L (SAAR191122LC)

storage temp.

2-30°C

Application

For laboratory use only

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service