Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Q100F

Sartobind® Membrane Adsorber Unit

matrix Q-type exchanger

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900

description

Q100
for ion exchange chromatography of biomolecules

feature

reusable

packaging

pkg of 1 unit

manufacturer/tradename

Sartorius

matrix

Q-type exchanger

Looking for similar products? Visit Product Comparison Guide

General description

Ready-to-use Sartobind membrane units are the ultimate for simple, ultra-rapid concentration of proteins from highly dilute solutions, or for separating proteins from a mixture. They feature high binding capacity (~1 mg/cm3), high flow rates (>100 mL/min.), and excellent resolution.

Use these units with syringes, peristaltic pumps, or preparative HPLC/FPLC workstations. the disposable 5 cm2 units are very useful for rapidly determining buffer conditions for ion exchange separations - simply connect units with different functionalities in series. Female Luer-Lock inlet/Male Luer-Lock outlet design.

Features and Benefits

Adsorption area: 100cm2

Legal Information

Sartobind is a registered trademark of Sartorius AG

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service