Skip to Content
MilliporeSigma
All Photos(1)

Documents

HABP02500

Millipore

MCE Membrane Filter, 0.45 μm Pore Size

MF-Millipore, filter diam. 25 mm, hydrophilic, black

Synonym(s):

MF-Millipore Membrane Filter, 0.45 µm pore size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

black filter
mixed cellulose esters (MCE) membrane
plain filter

Quality Level

description

25 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, black, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

parameter

4 L/min-cm2 air flow rate
60 mL/min-cm2 water flow rate
75 °C max. temp.

filter diam.

25 mm

thickness

150 μm

gravimetric extractables

2.5%

color

black

matrix

MF-Millipore

pore size

0.45 μm pore size
79 % porosity

capacity

150 μg/cm2 binding capacity (protein)

bubble point

≥2.2 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Application

  • Fluorescent bacteriological assays
  • Particle monitoring
  • Bioassays
  • Particle monitoring
  • Microplastics analysis grade water

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service