Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N11203

Bifenthrin

analytical standard

Synonym(s):

3-[(1Z)-2-Chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethylcyclopropanecarboxylic acid (2-methylbiphenyl-3-yl)methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22ClF3O2
CAS Number:
Molecular Weight:
422.87
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-2003

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

CC1=C(COC([C@@H]2[C@H](/C=C(Cl)/C(F)(F)F)C2(C)C)=O)C=CC=C1C3=CC=CC=C3

InChI

1S/C23H22ClF3O2/c1-14-16(10-7-11-17(14)15-8-5-4-6-9-15)13-29-21(28)20-18(22(20,2)3)12-19(24)23(25,26)27/h4-12,18,20H,13H2,1-3H3/b19-12-/t18-,20-/m0/s1

InChI key

OMFRMAHOUUJSGP-IRHGGOMRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1 - STOT RE 1

Target Organs

Nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

329.0 °F - closed cup

Flash Point(C)

165 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mayada R Farag et al.
Aquatic toxicology (Amsterdam, Netherlands), 231, 105715-105715 (2020-12-21)
This study was conceptualized in order to assess the 96-h LC50 of bifenthrin (BF) in O. niloticus and also to measure the biochemical, behavioral, and molecular responses of the fish suchronically exposed to a sub-lethal concentration of the insecticide. The
Amelie Segarra et al.
Toxics, 9(2) (2021-03-07)
Salinity can interact with organic compounds and modulate their toxicity. Studies have shown that the fraction of pyrethroid insecticides in the aqueous phase increases with increasing salinity, potentially increasing the risk of exposure for aquatic organisms at higher salinities. In
Weiying Jiang et al.
Environmental science & technology, 46(11), 6028-6034 (2012-05-11)
Pesticides such as pyrethroids have been frequently found in runoff water from urban areas and the offsite movement is a significant cause for aquatic toxicities in urban streams and estuaries. To better understand the origination of pesticide residues in urban
Amanda D Harwood et al.
Chemosphere, 90(3), 1117-1122 (2012-10-23)
Recent studies have determined that techniques, such as solid phase microextraction (SPME) fibers and Tenax beads, can predict bioaccumulation and potentially could predict toxicity for several compounds and species. Toxicity of bifenthrin was determined using two standard sediment toxicity tests
Chatchai Tananchai et al.
Journal of vector ecology : journal of the Society for Vector Ecology, 37(1), 187-196 (2012-05-03)
Contact and noncontact behavioral actions of wild-caught Anopheles dirus in response to the operational field dose of three synthetic pyrethroids (bifenthrin, α-cypermethrin and λ-cyhalothrin) were evaluated using an exito-repellency test chamber. DEET was used as the repellency standard for comparison

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service