MilliporeSigma
  • Neurobehavioral, physiological and inflammatory impairments in response to bifenthrin intoxication in Oreochromis niloticus fish: Role of dietary supplementation with Petroselinum crispum essential oil.
Neurobehavioral, physiological and inflammatory impairments in response to bifenthrin intoxication in Oreochromis niloticus fish: Role of dietary supplementation with Petroselinum crispum essential oil.
Aquatic toxicology (Amsterdam, Netherlands) (2020-12-21)
Mayada R Farag, Hemat K Mahmoud, Sabry A A El-Sayed, Sarah Y A Ahmed, Mahmoud Alagawany, Shimaa M Abou-Zeid
ABSTRACT
This study was conceptualized in order to assess the 96-h LC50 of bifenthrin (BF) in O. niloticus and also to measure the biochemical, behavioral, and molecular responses of the fish suchronically exposed to a sub-lethal concentration of the insecticide. The role of Petroselinum crispum essential oil (PEO) supplementation in mitigating the resulted neurotoxic insult was also investigated. The acute toxicity study revealed that the 96-h LC50 of BF is 6.81 μg/L, and varying degrees of behavioral changes were recorded in a dose-dependent manner. The subchronic study revealed reduction of dissolved oxygen and increased ammonia in aquaria of BF-exposed fish. Clinical signs revealed high degree of discomfort and aggressiveness together with reductions in survival rate and body weight gain. The levels of monoamines in brain, and GABA and amino acids in serum were reduced, together with decreased activities of Na+/K+-ATPase and acetylcholine esterases (AchE). The activities of antioxidant enzymes were also diminshed in the brain while oxdative damage and DNA breaks were elevated. Myeloperoxidase (MPO) activity in serum increased with overexpression of the pro-inflammatory cytokines in the brain tissue. BF also upregulated the expression of brain-stress related genes HSP70, Caspase-3 and P53. Supplemention of PEO to BF markedly abrogated the toxic impacts of the insecticide, specially at the high level. These findings demonstrate neuroprotective, antioxidant, genoprotective, anti-inflammatory and antiapoptic effects of PEO in BF-intoxicated fish. Based on these mechanistic insights of PEO, we recommend its use as an invaluable supplement in the fish feed.