Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K08670369

Kromasil® C8 HPLC Column

5 μm particle size, pore size 100 Å, L × I.D. 250 mm × 4.6 mm

Synonym(s):

C8 Reversed-Phase Chromatography Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52
Product K08670369 is not currently sold in your country. Contact Technical Service

product line

Kromasil®

manufacturer/tradename

Kromasil® KR100-5-C8-4.6X250

availability

available only in USA, Canada and Puerto Rico
available only in USA, Canada and Puerto Rico

technique(s)

HPLC: suitable

L × I.D.

250 mm × 4.6 mm

matrix active group

C8 (octyl) phase

particle size

5 μm

pore size

100 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Application

<ul>

<li><strong>Use of an UHPLC-MS/MS Method for Determination of Kuraridin and Characterization of Its Metabolites in Rat Plasma after Oral Administration.</strong>: This study developed and validated an ultra-high-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) method for determining Kuraridin and its metabolites in rat plasma. A C18 guard cartridge was employed to enhance the method′s efficacy and precision, demonstrating the cartridge′s utility in complex biological sample analysis. (Liu et al., 2018).</li>

<li><strong>Rapid and economical high-performance liquid chromatographic method for the determination of norfloxacin in serum using a microparticulate C18 guard cartridge.</strong>: This paper presents a cost-effective high-performance liquid chromatography (HPLC) method utilizing a microparticulate C18 guard cartridge for the rapid determination of norfloxacin in serum. The study highlights the cartridge′s ability to maintain high sensitivity and specificity while reducing analysis time and cost. (Wallis et al., 1995).</li>

</ul>

Legal Information

Kromasil is a registered trademark of EKA Chemicals AB

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What are the maximum temperature and pressure ratings for the Kromasil C8 HPLC Column, part number K08670369?

    1 answer
    1. Maximum operating pressure = 400 bar ( ≈ 6000 psi)<br>Maximum temperature = 60°C

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service