Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K08670482

Kromasil® Silica HPLC Column

5 μm particle size, pore size 60 Å, L × I.D. 250 mm × 4.6 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,150.00

$1,150.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$1,150.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

$1,150.00


Please contact Customer Service for Availability

Agency

suitable for USP L3

product line

Kromasil®

manufacturer/tradename

Kromasil® KR60-5-SIL-4.6X250

technique(s)

HPLC: suitable

L × I.D.

250 mm × 4.6 mm

matrix active group

silica phase

particle size

5 μm

pore size

60 Å

application(s)

food and beverages

separation technique

hydrophilic interaction (HILIC)
normal phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Kromasil is a registered trademark of EKA Chemicals AB

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the best way to store and clean out a Kromasil 100-5-C8 column? Thank you!

    1 answer
    1. Wash out all water and buffers from bonded silica columns and store in organic solvent. Unbonded silica columns may be stored in hexane, heptane or similar organic solvent. Keep in a cool area and replace end-caps to prevent the packing bed drying out.

      Please access the provided link for information on Care & Use of Kromasil HPLC Columns :https://www.kromasil.com/support/care_and_use.php

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service