Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM40605

Supelco

IPDI-DBA Oligomers Standard

certified reference material, in methanol (varied conc.), ampule of 1 mL

Synonym(s):

Isophorone diisocyanate dibutylamine oligomers standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

packaging

ampule of 1 mL

concentration

in methanol (varied conc.)

application(s)

environmental

format

single component solution

storage temp.

2-8°C

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    IPDI-Monomer-di-n-butylamine (isomer 1)
    IPDI-Monomer-di-n-butylamine (isomer 2)
    IPDI-isocyanurate-di-n-buyltamines (oligomers)

sample collection

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

121.1 °F - closed cup

Flash Point(C)

49.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service