Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5047-U

Supelco

ASSET® EZ4-NCO Dry Sampler

pkg of 2 ea

Synonym(s):

ASSET® EZ4 Dry Samplers

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$197.00

$197.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 PKG
$197.00

About This Item

UNSPSC Code:
12190000
NACRES:
NB.24

$197.00


Please contact Customer Service for Availability

Request a Bulk Order

description

Kit (each kit contains: 2 sampling cartridges, 2 tubing adapters, 1 calibration fitting and datasheet)

Quality Level

product line

ASSET®

packaging

pkg of 2 ea

technique(s)

active air sampling: suitable

matrix

DBA-impregnanated glass fiber filter

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for sampling isocyanate monomers & oligomers (*not recommended for isocyanic acid (ICA))
for use with LC-MS or LC-MS/MS

Looking for similar products? Visit Product Comparison Guide

General description

We offer a "try me size" ASSET EZ4-NCO dry sampler kit that contains 2 ASSET EZ4-NCO cartridges, 2 tubing adapters, and 1 calibration fitting. The kit makes it easy to get started sampling with ASSET - 1 cartridge for sampling and the other as a field blank.

Legal Information

Asset is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B Inhalation - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Gylestam et al.
The Annals of occupational hygiene, 58(3), 340-354 (2013-12-29)
An advanced design of a denuder impactor (DI) sampler has been developed for characterization of possible airborne isocyanate exposure in different particle size fractions. The sampler is equipped with 12 different parallel denuder tubes, 4 impaction stages with the cut-off

Related Content

Easy-to-use air sampler offering ultimate sensitivity for collection and measurement of vapor phase and particulate isocyanate monomers and oligomers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service