Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

843272

TOYOPEARL® SP-550C Bulk Media

phase sulfopropyl, bottle of 100 mL, 100 μm particle size, Tosoh 0043272

Synonym(s):

TOYOPEARL® SP-550C Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

product line

TOYOPEARL®

form

slurry

feature

counter ion Na+

packaging

bottle of 100 mL

manufacturer/tradename

Tosoh 0043272
Tosoh 43272

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

sulfopropyl phase

particle size

100 μm

operating pH

2-13

capacity

100 g/L±20 adsorption capacity (lysozyme)

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl SP-550 resin is a strong cation exchanger useful for purification of molecules such as insulin and proteins up to 100kD. It is used for high throughput capture of feedstocks.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service