Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

814028

Supelco

TOYOPEARL® SP-550C Bulk Media

phase sulfopropyl, bottle of 250 mL, 100 μm particle size, Tosoh 0014028

Synonym(s):

TOYOPEARL® SP-550C Bulk Media

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$697.00

$697.00


Please contact Customer Service for Availability


Select a Size

Change View
250 ML
$697.00

About This Item

UNSPSC Code:
47101511

$697.00


Please contact Customer Service for Availability

description

SP-550C

product line

TOYOPEARL®

form

slurry

feature

counter ion Na+

packaging

bottle of 250 mL

manufacturer/tradename

Tosoh 0014028

parameter

3 bar max. pressure

technique(s)

HPLC: suitable

matrix

methacrylate

matrix active group

sulfopropyl phase

particle size

100 μm

operating pH

3-11

capacity

0.15 meq/mL±0.02 meq/mL ion exchange capacity
100 g/L±20 adsorption capacity (lysozyme)
111 mg/mL binding capacity (bovine serum albumin)

compatibility

mode of use strong cation exchange chromatography

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl SP-550 resin is a strong cation exchanger useful for purification of molecules such as insulin and proteins up to 100kD. It is used for high throughput capture of feedstocks.

Application

TOYOPEARL® SP-550C columns are used in separation media and ion exchange resins.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arne Staby et al.
Journal of chromatography. A, 1034(1-2), 85-97 (2004-05-01)
A comparative study was performed on strong cation-exchangers to investigate the pH dependence, efficiency, binding strength, particle size distribution, static and dynamic capacity, and SEM pictures of chromatographic resins. The resins tested included: SP Sepharose XL, Poros 50 HS, Toyopearl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service