Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82034-U

Hamilton PRP-X300 HPLC Guard Cartridge

Kit, 12 μm particle size, L × I.D. 2 mm × 2.1 mm, stainless steel hardware

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

material

stainless steel hardware

Agency

suitable for USP L17

packaging

pkg of 1 kit

technique(s)

HPLC: suitable

L × I.D.

2 mm × 2.1 mm

matrix

PS-DVB particle platform
polymeric

matrix active group

sulfonic acid phase

particle size

12 μm

pore size

100 Å

operating pH

1-14

separation technique

ion exclusion

Looking for similar products? Visit Product Comparison Guide

General description

Kit includes 1 holder and 2 cartridges

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service