Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

818001

TSKgel® SuperHM-H (3 µm) HPLC Columns

L x I.D. 15 cm × 6 mm, polystyrene-divinylbenzene phase

Synonym(s):

TSKgel® SuperH Columns

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$3,060.00

$3,060.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$3,060.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

$3,060.00


Please contact Customer Service for Availability

Product Name

TSKgel® SuperHM-H HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 15 cm × 6 mm, 3 μm particle size

material

stainless steel hardware

Agency

suitable for USP L21

product line

TSKgel®

manufacturer/tradename

Tosoh 18001

parameter

0.8 mL/min flow rate
140 °C temperature
39 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

15 cm × 6 mm

matrix

polymer particle platform

matrix active group

polystyrene-divinylbenzene phase

particle size

3 μm

pore size

mixed

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel SuperHM-H mixed-bed columns are packed with rigid polymer beads with varying pore sizes. They are used for the non-aqueous separation of polymers with a broad molecular weight range or for analyzing unknown samples. TSKgel SuperH columns are shipped in THF, which can be exchanged for a wide variety of organic solvents.
The TSKgel SuperH column line consists of eight columns with different pore sizes, TSKgel SuperH1000 through TSKgel SuperH7000, and four columns with an extended linear range of the calibration curve. TSKgel SuperH-type columns are high efficiency/high throughput versions of the conventional TSKgel HHR columns. Both column types are based on the same bead chemistry. Maximum operating temperature for TSKgel SuperH-type columns is 140°C. All TSKgel SuperH-type columns are shipped in THF, which can be exchanged for a wide variety of organic solvents. TSKgel SuperH-type columns are high efficiency, high-throughput gel permeation columns with dimensions of 6mm ID x 15cm.

Physical form

Shipped in THF.

Other Notes

THF can be replaced with acetone, ethanol, quinoline, benzene, o-dichlorobenzene, ethyl acetate, dodecane, FC-113, carbon tetrachloride, dichloromethane, dichloroethane, trichloroethane, n-hexane, cyclohexane, xylene, tetrahydrofuran, chloroform, 1,4-dioxane, hexafluoroisopropanol, toluene, 1-chloronaphthalene, dimethylaceto acetamide, methyl ethyl ketone, trichlorobenzene, m-cresol, dimethylformamide, methylpyrrolidone, o-chlorophenol/chloroform, dimethyl sulfoxide, pyridine

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service