Skip to Content
MilliporeSigma
vibrant-m

TSKgel® SuperHM-H (3 µm) HPLC Columns

vibrant-m
Synonym(s):
TSKgel® SuperH Columns

Products

Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1