Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

816173

TOYOPEARL® Ether-650M Bulk Media

matrix polymer (65 μm), bottle of 100 mL, 100 μm

Synonym(s):

TOYOPEARL® Ether-650 HIC Bulk Media

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$783.00

$783.00


Please contact Customer Service for Availability


Select a Size

Change View
100 ML
$783.00

About This Item

UNSPSC Code:
12000000

$783.00


Please contact Customer Service for Availability

description

Ether-650M

product line

TOYOPEARL®

form

liquid

packaging

bottle of 100 mL

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

polymer (65 μm)

matrix active group

Ether phase

particle size

100 μm

operating pH

1-13

separation technique

hydrophobic interaction (HIC)

General description

Toyopearl Ether-650 resin has the highest hydrophilicity of the HIC ligands offered by Tosoh Bioscience. It is useful for very hydrophobic molecules such as membrane proteins. It is typically used in intermediate purification and polishing steps. For high performance separations, the use of TSKgel Ether-5PW resin is suggested. Both products have the same selectivity.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service