Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

814698

TOYOPEARL® SP-650S Bulk Media

bottle of 1 L, phase sulphonyl, 35 μm particle size

Synonym(s):

TOYOPEARL® SP-650 Cation Exchange Media

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$6,970.00

$6,970.00


Please contact Customer Service for Availability


Select a Size

Change View
1 L
$6,970.00

About This Item

UNSPSC Code:
41115700

$6,970.00


Please contact Customer Service for Availability

product line

TOYOPEARL®

form

slurry

feature

counter ion Na+

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 14698

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

sulphonyl phase

particle size

35 μm

operating pH

2-13

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl SP-650 resin is a strong cation exchanger for biomolecule purification. It can be used for high throughput capture, intermediate purification, and polishing process steps. If more powerful resolution is needed the smaller particle TSKgel SP-5PW resin should be used. Selectivity remains the same.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service