Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808805

TSKgel® SW Type (10 µm) HPLC Column

L x I.D. 4 cm × 8 mm, diol phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$977.00

$977.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$977.00

About This Item

UNSPSC Code:
41115711

$977.00


Please contact Customer Service for Availability

Product Name

TSKgel® SW-Type Guard Column, phase diol, L × I.D. 4 cm × 8 mm, 10 μm particle size, glass

material

glass

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 08805

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
0.8 mL/min flow rate
10-30 °C temperature

technique(s)

HPLC: suitable

L × I.D.

4 cm × 8 mm

matrix

spherical silica

matrix active group

diol phase

particle size

10 μm

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

General description

Guard Column for 30 and 60 cm x 7.5mmID TSKgel® G2000SW, G3000SW, G4000SW columns.

Physical form

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service