Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808095

Endfitting with fixed 2 μm frit for all 7.8 mm I.D. stainless steel TSKgel® Columns

stainless steel, pkg of 1 ea

Synonym(s):

TSKgel® Hardware and Accessories

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$173.00

$173.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$173.00

About This Item

UNSPSC Code:
41115700

$173.00


Please contact Customer Service for Availability

material

stainless steel

product line

TSKgel®

packaging

pkg of 1 ea

manufacturer/tradename

Tosoh 08095

technique(s)

HPLC: suitable

pore size

2 μm

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service