Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

807619

TSKgel® HPLC Columns Hardware and Accessories

pkg of 1 ea

Synonym(s):

Endfitting with fixed 2 μm frit for all 4.6 mm I.D. TSKgel® stainless steel columns

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$173.00

$173.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$173.00

About This Item

UNSPSC Code:
41115700

$173.00


Please contact Customer Service for Availability

Product Name

TSKgel® Hardware and Accessories, pkg of 1 ea

material

stainless steel

product line

TSKgel®

packaging

pkg of 1 ea

manufacturer/tradename

Tosoh 07619

technique(s)

HPLC: suitable

pore size

2 μm

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service