Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

807472

TOYOPEARL® DEAE-650S Bulk Media

phase DEAE (diethylaminoethyl), bottle of 250 mL, 35 μm particle size

Synonym(s):

TOYOPEARL® DEAE-650C Bulk Media

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$1,980.00

$1,980.00


Please contact Customer Service for Availability


Select a Size

Change View
250 ML
$1,980.00

About This Item

UNSPSC Code:
47101511

$1,980.00


Please contact Customer Service for Availability

description

DEAE-650S

product line

TOYOPEARL®

form

slurry

feature

counter ion Cl-

packaging

bottle of 250 mL

parameter

3 bar max. pressure

technique(s)

HPLC: suitable

matrix

methacrylate

matrix active group

DEAE (diethylaminoethyl) phase

particle size

35 μm

operating pH

2-10

capacity

0.10 meq/mL±0.02 meq/mL ion exchange capacity
30 g/L±5 adsorption capacity (BSA)
30 mg/mL±5 mg/mL binding capacity (bovine serum albumin)

compatibility

mode of use weak anion exchange chromatography

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl DEAE-650 resin is a weak anion exchanger for blood fractionation and other biomolecule purification. It can be used for high throughput capture, intermediate purification, and polishing process steps. If more powerful resolution is needed TSKgel DEAE-5PW resin should be used. Selectivity remains the same.

Application

TOYOPEARL® DEAE-650S columns are used as separation media and ion exchange resins.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafael Picorel et al.
Methods in molecular biology (Clifton, N.J.), 274, 129-135 (2004-06-10)
A single-column method to purify the CP43 and CP47 pigment-protein complexes of photo-system (PS)II from higher plants is presented. To validate the isolation procedure, three different species were used (Spinacea oleracea, Beta vulgaris, and Glycine max), and the procedure worked
Rafael Picorel et al.
Methods in molecular biology (Clifton, N.J.), 684, 105-112 (2010-10-21)
A method to isolate and purify CP43 and CP47 pigment-protein complexes from Photosystem (PS) II of higher plants is presented. The method has been developed in spinach, but it may also be valid for other plant species, since there is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service