Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

807168

TSKgel® DEAE-SW Columns

phase DEAE (diethylaminoethyl), L × I.D. 25 cm × 4.6 mm, 5 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
Pricing and availability is not currently available.

feature

counter ion H2PO4-

manufacturer/tradename

Tosoh 07168

parameter

≤1 M salt concentration
0-100% organic solvent
1.0 mL/min flow rate
10-45 °C temperature
150 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

matrix

silica particle platform

matrix active group

DEAE (diethylaminoethyl) phase

particle size

5 μm

pore size

125 Å

operating pH

2.0-7.5

separation technique

anion exchange

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel DEAE-2SW columns are packed with 5μm porous spherical silica beads which are surface modified with a weak anion exchanger. They are used for the separation and analysis of nucleotides, nucleosides, peptides, small proteins and other biologically active molecules. A 2mm ID column is available for applications that are sample limited.

Physical form

Shipped in methanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service