Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58743

Tubing Adapter

1/4-28 male UNF, for 1/8 in. tube

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$50.80

$50.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$50.80

About This Item

UNSPSC Code:
41115700

$50.80


Please contact Customer Service for Availability

Request a Bulk Order

material

stainless steel unit

description

for chromatography applications

packaging

pkg of 1 ea

bore

1.5 mm (0.60 in.)

fitting

for 1/8 in. tube
1/4-28 male UNF

Looking for similar products? Visit Product Comparison Guide

General description

Tubing Adapter is an accessory that can be used with low pressure liquid chromatography (LPLC) and flash chromatography columns.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Valco is a registered trademark of Valco Instruments Co. Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service