Key Documents
57632
Sample Loop for Model 7725 Injector
volume 5 mL
About This Item
UNSPSC Code:
41115854
Pricing and availability is not currently available.
volume
5 mL
General description
Sample loop for model 7725 injector (volume 5 μL) is fabricated for advanced fluid-handling solutions for large number of analytical instrumentation and in vitro diagnostic systems. It can be made use in high performance liquid chromatographic (HPLC) systems and is quite accurate and precise in injecting 1 μL-5 mL concentration of samples.
Other Notes
Note: Use VESPEL seals to pH 10, Tefzel seals to pH 14.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)