Skip to Content
MilliporeSigma
All Photos(1)

Documents

57649

Sample Loop for 9010, 9125, and 9725 Injectors

volume 5 mL, PEEK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

material

PEEK

O.D.

1.6 mm
1/16 in.

volume

5 mL

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service