Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

568420-U

Supelco

Discovery® BIO Wide Pore C5 (5 µm) HPLC Columns

L × I.D. 5 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Discovery® BIO Wide Pore C5 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm

material

stainless steel column

product line

Discovery®

feature

endcapped

manufacturer/tradename

Discovery®

packaging

1 ea of

extent of labeling

3.5% Carbon loading

parameter

0-70 °C temperature
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

5 cm × 4.6 mm

surface area

100 m2/g

surface coverage

4.5 μmol/m2

matrix

silica particle platform
fully porous particle

matrix active group

C5 (pentyl) phase

particle size

5 μm

pore size

300 Å

operating pH

2-8

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Discovery BIO Wide Pore C5 was designed for the efficient and reliable separation of proteins and peptides, especially hydrophobic peptides, by RP- HPLC. Long-chain phases, like C8 or C18, are often too hydrophobic for proteins and can cause excessively long retention time or even irreversible binding to the column. For this reason short-chain phases, typically C3 or C4, are often used for RP-HPLC of proteins. However, these short-chain phases are susceptible to hydrolysis, resulting in short column lifetime, especially at low pH. The Discovery BIO Wide Pore C5 gives elution order similar to a conventional C4, yet has enhanced pH stability for longer column lifetime. Generally, higher efficiency separations are achievable on the Discovery BIO Wide Pore C5 because of the improvements we have made to the silica and bonded-phase chemistry.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Discovery BIO Wide Pore C5 products offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service