Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55079

Upchurch Biocompatible Precolumn Filter

pore size 0.5 μm, pkg of 1 ea

Synonym(s):

HIGH PRESSURE INLINE FILTER, 0.5UM, EA

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$45.70

$45.70


Available to ship onApril 14, 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$45.70

About This Item

UNSPSC Code:
41115700

$45.70


Available to ship onApril 14, 2025Details


Request a Bulk Order

material

PEEK frit and body
stainless steel holder

packaging

pkg of 1 ea

manufacturer/tradename

A-431
Upchurch Scientific, Inc

pore size

0.5 μm

Looking for similar products? Visit Product Comparison Guide

General description

In-line installation. Stainless steel body with inert polyetherether-ketone (PEEK) end fittings. Choose either a 0.5 μm or 2 μm PEEK frit in one endfitting.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.

Questions

  1. Just to confirm, #55079 contains the 0.5um frit/filter and the stainless steel housing? Thanks

    1 answer
    1. 55079 is made of a stainless steel body with an inert PEEK frit and body.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service