Key Documents
502707
PEEK Inline Microfilter
Select a Size
1 EA
$174.00
$174.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$174.00
About This Item
UNSPSC Code:
41115700
$174.00
Please contact Customer Service for Availability
Request a Bulk Order
packaging
pkg of 1 ea
General description
Install between 2 pieces of capillary tubing, using Microtight tubing sleeves. PEEK housing with special PEEK fitting that encases the 0.5 μm PEEK frit. Through hole 0.006 in., swept volume 0.24 μL. Includes 2 MicroFingertight nuts, and 5 frits (1 installed).
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)