509620
Syringes Fitted with Luer Lock Valve (509353)
volume 10 mL, 10MDR-VLLMA-GT, needle size (not included)
All Photos(1)
About This Item
Recommended Products
material
glass syringe
needle size
(not included)
feature
Pre-fitted with Luer Lock Valve
packaging
pkg of 1 ea
manufacturer/tradename
SGE 008970
volume
10 mL
Related Categories
General description
- SGE Syringes [volume 10 mL, needle size (not included)] fitted with Luer Lock Valve, Model: 10MDR-VLLMA-GT.
- Fabricated using borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
- Autoclavable syringes but the plunger removal is necessary.
- Have an interference fit between the plunger tip and the barrel that enhances the gas sealing properties of the syringe.
- Maximum inertness for liquid or gas samples.
Choose from one of the most recent versions:
Certificates of Analysis (COA)